missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DEFA1 (Human) Recombinant Protein
Human DEFA1 full-length ORF ( AAH27917, 1 a.a. - 94 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH27917 |
---|---|
Gene ID (Entrez) | 1667 |
Name | defensin, alpha 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16114001
|
Abnova™
H00001667-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16124001
|
Abnova™
H00001667-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MRTLAILAAILLVALQAQAEPLQARADEVAAAPEQIAADIPEVVVSLAWDESLAPKHPGSRKSMDCYCRIPACIAGERRYGTCIYQGRLWAFCC
Specifications
AAH27917 | |
defensin, alpha 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
DEF1, DEFA2, HNP-1, HP-1, MGC138393, MRS | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1667 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEFA1 | |
GST |