Description
- Centrifuge bucket
- For rotor A-2-DWP (CFA-154-010B) deepwel
- Available in packs of 2
Specifications
Specifications
| For Use With (Equipment) | Eppendorf rotor CFA-154-010B (A-2-DWP) |
| Quantity | 2/Pk. |