missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin-2/CLEC6A Antibody (545925), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB31141H
This item is not returnable.
View return policy
Description
Dectin-2/CLEC6A Monoclonal antibody specifically detects Dectin-2/CLEC6A in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Dectin-2/CLEC6A | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 545925 | |
| Flow Cytometry, CyTOF-ready | |
| CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
| Mouse myeloma cell line NS0-derived recombinant human Dectin-2/CLEC6A, Thr46-Leu209, Accession # Q6EIG7 | |
| RUO | |
| 93978 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction