Learn More
Thermo Scientific™ Deactivated Glass Wool
Product Code. p-9458002 Shop All Thermo Scientific Products
Click to view available options
Description:
Deactivated Glass Wool 10 g
Unit Size:
Each
Product Code. 11567781
Brand: Thermo Scientific™ 60180785
Only null left
Add to basket
Add to basket
Pack GC injection port liners with deactivated glass wool for good homogenization, volatilization of high boiling point compounds, and minimal column contamination.
Pack your GC injection port liner with Thermo Scientific™ deactivated glass wool for good homogenization and volatilization of high boiling point compounds within the liner. The glass wool acts as a barrier to prevent non-volatile sample matrix compounds and septa particles from entering GC columns, minimizing contamination and increasing precision and reproducibility.
TRUSTED_SUSTAINABILITY
Specifications
Description Deactivated Glass Wool 10 g
Quantity 10 g
Packing Material Deactivated Glass Wool
For Use With (Equipment) GC Liners