missing translation for 'onlineSavingsMsg'
Learn More

DDX39 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18555623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18555623 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18555623 Supplier Novus Biologicals Supplier No. NBP298622AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDX39 Polyclonal antibody specifically detects DDX39 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias ATP-dependent RNA helicase DDX39, BAT1, BAT1L, DDX39, DDXL, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 transcript, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A, DEAD box protein 39, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39, EC 3.6.4.13, MGC18203, MGC8417, Nuclear RNA helicase URH49, nuclear RNA helicase, DECD variant of DEAD box family, UAP56-related helicase, 49 kDa, URH49
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DDX39 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10212
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.