missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDT Antibody (OTI1H3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70562PE
This item is not returnable.
View return policy
Description
DDT Monoclonal antibody specifically detects DDT in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| DDT | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1H3 | |
| Western Blot, Immunohistochemistry | |
| D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II | |
| Full length human recombinant protein of human DDT(NP_001346) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1652 | |
| Store at 4C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion