missing translation for 'onlineSavingsMsg'
Learn More

DDR2 Antibody (9A9), Novus Biologicals™

Rabbit Monoclonal Antibody

355.00€ - 533.00€

Specifications

Antigen DDR2
Clone 9A9
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043317
View Documents
Novus Biologicals
NBP3-26628-50ul
50 μL
355.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043897
View Documents
Novus Biologicals
NBP3-26628-100ul
100 μL
533.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DDR2 Monoclonal antibody specifically detects DDR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications

Specifications

DDR2
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Monoclonal
Purified
RUO
Human, Mouse
CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase
A synthesized peptide derived from Human DDR2 [UniProt Q16832]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
9A9
Western Blot, Flow Cytometry, ELISA
Unconjugated
Rabbit
Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction
PBS, pH 7.4, 150mM NaCl and 50% glycerol
4921
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.