Learn More
DDB1 Mouse, anti-Human, CoraLite Plus 488, Clone: 4F6C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66010-20UL
Description
The DDB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDB1. This antibody recognizes human, mouse, rat antigen. The DDB1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| DDB1 | |
| 4F6C6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG2b |
| Western Blot | |
| 1000 μg/mL | |
| DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor | |
| 20 μL | |
| 1642 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |