Learn More
DDB1 Mouse, anti-Human, CoraLite Plus 488, Clone: 4F6C6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66010-20UL
 View more versions of this product
Product Code. 18050316
  • 145.00€ / 20µL
Description
The DDB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DDB1. This antibody recognizes human, mouse, rat antigen. The DDB1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
DDB1
4F6C6
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG2b
Western Blot
1000 μg/mL
DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor
20 μL
1642
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.