missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCIR/CLEC4A Antibody (9E8) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | DCIR/CLEC4A |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 9E8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunising BALB/c mice with a dendritic cell immunoreceptor ectodomain-human Fc fusion protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?