missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam DCIA, Fluorescent Thiol-reactive Neutral Probe, Solid, 100 mg
Description
- Chemical Properties: Fluorescent thiol-reactive neutral probe that is self-quenching until it reacts with thiol groups, at which point it regains strong fluorescence.
- Molecular Weight: 490.33 g/mol
- Purity: >85%
- Physical Form: Solid
- Chemical Formula Components: 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin
- CAS Number: 76877-34-4
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | Thiol groups |
| Target Molecule | Thiol groups |
| Detection Method | Fluorescence |
| Formulation | 100% 7-Diethylamino-3-[4-(iodoacetamido)phenyl]-4-methylcoumarin |
| Quantity | 100 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?