missing translation for 'onlineSavingsMsg'
Learn More

DC-SIGN/CD209 Antibody (120507) [DyLight 755], Novus Biologicals™

Product Code. 18533956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18533956 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18533956 Supplier Novus Biologicals Supplier No. FAB161Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC-SIGN/CD209
Applications Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF
Classification Monoclonal
Clone 120507
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Host Species Mouse
Immunogen NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGN/CD209
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 30835
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.