missing translation for 'onlineSavingsMsg'
Learn More

DBeQ, Tocris Bioscience™

Product Code. 18094252
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094252 50 mg 50mg
18035342 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094252 Supplier Tocris Bioscience Supplier No. 4417/50

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Selective and reversible p97 ATPase inhibitor

Selective and reversible inhibitor of p97 ATPase (VCP, IC50 = 1.5 μM). Induces executioner caspases (caspase-3 and caspase-7) rapidly. Blocks the degradation of endoplasmic reticulum-associated degradation (ERAD) reporters; also blocks autophagosome maturation and promotes accumulation of LC3-II.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177355-84-9
Molecular Formula C22H20N4
Molecular Weight (g/mol) 340.43
MDL Number MFCD03691820
InChI Key QAIMUUJJAJBPCL-UHFFFAOYSA-N
Synonym dbeq, n2,n4-dibenzylquinazoline-2,4-diamine, n,n'-dibenzylquinazoline-2,4-diamine, 2,4-dibencilaminoquinazoline, 2,4-quinazolinediamine, n,n'-bis phenylmethyl, n*2*,n*4*-dibenzyl-quinazoline-2,4-diamine, n2,n4-bis phenylmethyl-2,4-quinazolinediamine, jrf12, jrf 12, jrf-12
PubChem CID 676352
IUPAC Name N2,N4-dibenzylquinazoline-2,4-diamine
SMILES C(NC1=NC(NCC2=CC=CC=C2)=C2C=CC=CC2=N1)C1=CC=CC=C1

Specifications

Quantity 50 mg
Formula Weight 340.42
Percent Purity >99%
Chemical Name or Material DBeQ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.