accessibility menu, dialog, popup

UserName

CAS RN 177355-84-9

HNNNHN

CAS RN 177355-84-9

IUPAC Name: N2,N4-dibenzylquinazoline-2,4-diamine
Synonyms: N2,N4-dibenzylquinazoline-2,4-diamine
Molecular Weight (g/mol): 340.43
Molecular Formula: C22H20N4
InChi Key: QAIMUUJJAJBPCL-UHFFFAOYSA-N
SMILES: C(NC1=NC(NCC2=CC=CC=C2)=C2C=CC=CC2=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results