missing translation for 'onlineSavingsMsg'
Learn More

DAZAP2 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18632701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18632701 0.02 mL 0.02mL
18627800 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18632701 Supplier Novus Biologicals Supplier No. NBP2921440.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DAZAP2 Polyclonal antibody specifically detects DAZAP2 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAZAP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DAZ associated protein 2, deleted in azoospermia associated protein 2, Deleted in azoospermia-associated protein 2, KIAA0058DAZ-associated protein 2, MGC14319, MGC766, proline-rich transcript in brain, proline-rich transcript, brain-expressed protein, PRTB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-168 of human DAZAP2 (NP_055579.1). MNSKGQYPTQPTYPVQPPGNPVYPQTLHLPQAPPYTDAPPAYSELYRPSFVHPGAATVPTMSAAFPGASLYLPMAQSVAVGPLGSTIPMAYYPVGPIYPPGSTVLVEGGYDAGARFGAGATAGNIPPPPPGCPPNAAQLAVMQGANVLVTQRKGNFFMGGSDGGYTIW
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9802
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.