missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15793-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein which is part of mitochondrial complex 1, part of the oxidative phosphorylation system in mitochondria. Complex 1 transfers electrons to ubiquinone from NADH which establishes a proton gradient for the generation of ATP. Mutations in this gene are associated with Leigh syndrome due to mitochondrial complex 1 deficiency. Pseudogenes of this gene are located on chromosomes 5 and 13. Alternative splicing results in multiple transcript variants.Specifications
| DAP13 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA12 | |
| B17.2, CI B17.2, CIB17.2, Complex I B17.2, DAP13, NDUFA12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 299739, 55967, 66414 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TMF3, Q9UI09 | |
| NDUFA12 | |
| DAP13 Fusion Protein Ag8501 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction