missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP Kinase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
167.00€
Specifications
| Antigen | DAP Kinase 2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
DAP Kinase 2 Polyclonal specifically detects DAP Kinase 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DAP Kinase 2 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Autophagy, Cellular Markers | |
| PBS with 0.02% Sodium Azide | |
| DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1 | |
| DAPK2 | |
| IgG | |
| Affinity Purified | |
| DAPK2 has no cross responses to DAPK1. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| BAA88063 | |
| 23604 | |
| Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human DAPK2. This antigenic peptide is identical to the corresponding amino acids of mouse origin (1,2). The immunogen is located within the last 50 amino acids of DAPK2. Amino Acid Squence: ARRKALHPRRRSSTS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title