Learn More
Dacarbazine
Product Code. 15463689
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463689 500 mg 500mg
15453689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15463689 Supplier Thermo Scientific Alfa Aesar Supplier No. J61023.ME
 Restricted Item
In Stock
Add to basket
CAS: 4342-03-4 | C6H10N6O | 182.19 g/mol
Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Used in treatment of malignant melanoma and sarcomas
  • Generates methyl adducts to purine bases in DNA
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3-4-4342
MDL Number MFCD00057167
Specifications
Melting Point 210°C to 212°C
Color White to Yellow
Molecular Formula C4H3IN2O
Quantity 500 mg
Merck Index 14,2798
Synonym 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one
Solubility Information Slightly soluble in water,DMSO and ethanol /n
InChI Key SMIGOGPUTQXOKF-UHFFFAOYSA-N
SMILES C1=C(C(=O)NC=N1)I
IUPAC Name 5-iodo-1H-pyrimidin-6-one
Molecular Weight (g/mol) 221.99
PubChem CID 12356582
Formula Weight 182.18
Physical Form Powder
Chemical Name or Material Dacarbazine