Learn More
DAB2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10109-100UL
Description
The DAB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DAB2. This antibody recognizes human antigen. The DAB2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| DAB2 | |
| 1000 μg/mL | |
| DAB2, Disabled homolog 2, DOC 2, DOC2, FLJ26626 | |
| 100 μL | |
| 1601 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |