Learn More
DAB2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10109-100UL
 View more versions of this product
Product Code. 18068025
  • 450.00€ / 100µL
Description
The DAB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DAB2. This antibody recognizes human antigen. The DAB2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
DAB2
1000 μg/mL
DAB2, Disabled homolog 2, DOC 2, DOC2, FLJ26626
100 μL
1601
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa