missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAB2 Monoclonal Antibody (883216)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561902
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta).
Specifications
| DAB2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| TBS with trehalose and no preservative | |
| P98082 | |
| DAB2 | |
| E. coli-derived recombinant human DAB2, Lys630-Ala770. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 883216 | |
| Unconjugated | |
| DAB2 | |
| 5730435J12Rik; AA960054; Adaptor molecule disabled-2; AI957090; C9; D15Wsu122e; D630005B22Rik; Dab, mitogen-responsive phosphoprotein, homolog 2; Dab2; DAB2, clathrin adaptor protein; differentially expressed in ovarian carcinoma 2; Differentially-expressed protein 2; disabled 2, mitogen-responsive phosphoprotein; disabled homolog 2; disabled homolog 2 mitogen-responsive phosphoprotein; disabled homolog 2, mitogen-responsive phosphoprotein; Doc2; Doc-2; DOC-2 p82 isoform; FLJ26626; Mitogen-responsive phosphoprotein; p96 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction