missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAB2 Antibody (2H7C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
228.00€
Specifications
| Antigen | DAB2 |
|---|---|
| Clone | 2H7C4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Description
DAB2 Monoclonal specifically detects DAB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| DAB2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Differentially-expressed protein 2, disabled (Drosophila) homolog 2 (mitogen-responsive phosphoprotein), disabled homolog 2, mitogen-responsive phosphoprotein (Drosophila), DOC-2DOC2disabled homolog 2, FLJ26626, mitogen-responsive phosphoprotein | |
| DAB2 | |
| IgG1 | |
| Protein G purified |
| 2H7C4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| Neuroscience | |
| 1601 | |
| Purified recombinant fragment of human DOC2 (AA: 652-749) expressed in E. Coli. | |
| Primary | |
| 23.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title