missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DAB1 Antibody - N-terminal, Novus Biologicals™

Product Code. 30066953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066953 50 μg 50µg
30067034 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30066953 Supplier Novus Biologicals™ Supplier No. NBP33079450ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

DAB1 Polyclonal antibody specifically detects DAB1 in Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAB1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1-5 μg/mL, Immunoprecipitation : 5-10 μg/mL
Formulation Phosphate buffered saline
Gene Alias disabled (Drosophila) homolog 1, disabled homolog 1, disabled homolog 1 (Drosophila)
Host Species Goat
Immunogen Goats were immunized with a recombinant protein corresponding to amino acids 1-179 of mouse disabled coupled to a GST fusion protein.
Purification Method Ammonium sulfate precipitation
Quantity 50 μg
Regulatory Status RUO
Research Discipline Growth and Development, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1600
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.