Learn More
Abcam DAB Enhancer, Liquid, 10 mL
Product Code. 32118127
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32118127 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32118127 Supplier Abcam Supplier No. ab67510mL
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
DAB signal enhancer designed for use with peroxidase-based immunostaining systems.
  • Application: Use as a DAB signal enhancer in conjunction with peroxidase-based immunostaining systems.
  • Form: Liquid.
  • Storage: Store at ambient temperature.
  • Shipping: Ambient (cannot ship with ice).
  • Constituents: 100% 3,3'-Diaminobenzidine.
Specifications
Buffer 100% 3,3'-Diaminobenzidine
Content And Storage Store at ambient temperature.
Format Bottle
Form Liquid
Kit Contents DAB Enhancer, 10 mL
Reactivity Peroxidase
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate DAB
Target Molecule 3,3'-Diaminobenzidine
Detection Method Chromogenic
Formulation 100% 3,3'-Diaminobenzidine
Quantity 10 mL
For Use With (Application) Peroxidase-based immunostaining