missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D2HGDH Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 463.00€
Specifications
| Antigen | D2HGDH |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624971
|
Novus Biologicals
NBP2-92309-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607661
|
Novus Biologicals
NBP2-92309-0.1ml |
0.1 mL |
463.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D2HGDH Polyclonal antibody specifically detects D2HGDH in Mouse, Rat samples. It is validated for Western BlotSpecifications
| D2HGDH | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 728294 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| D-2-hydroxyglutarate dehydrogenase, EC 1.1.99, EC 1.1.99.-, FLJ42195, MGC25181, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 322-521 of human D2HGDH (NP_689996.4). GRHLHLASPVQESPFYVLIETSGSNAGHDAEKLGHFLEHALGSGLVTDGTMATDQRKVKMLWALRERITEALSRDGYVYKYDLSLPVERLYDIVTDLRARLGPHAKHVVGYGHLGDGNLHLNVTAEAFSPSLLAALEPHVYEWTAGQQGSVSAEHGVGFRKRDVLGYSKPPGALQLMQQLKALLDPKGILNPYKTLPSQA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title