Learn More
D-Galactal, TRC
Product Code. 30012772
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012772 5 g 5g
30006107 25 g 25g
30007461 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012772 Supplier Toronto Research Chemicals Supplier No. TRCG1510005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21193-75-9
Molecular Formula C6 H10 O4
Molecular Weight (g/mol) 146.14
IUPAC Name (2R,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
SMILES OC[C@H]1OC=C[C@@H](O)[C@H]1O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H10O4/c7-3-5-6(9)4(8)1-2-10-5/h1-2,4-9H,3H2/t4-,5-,6-/m1/s1
Formula Weight 146.0579
Chemical Name or Material D-Galactal