CAS RN 21193-75-9
IUPAC Name:
2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
Synonyms:
2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
Molecular Weight (g/mol):
146.14
Molecular Formula:
C6H10O4
InChi Key:
YVECGMZCTULTIS-UHFFFAOYNA-N
SMILES:
OCC1OC=CC(O)C1O
1
–
1
of
1
results
1
D-Galactal, TRC
CAS: 21193-75-9 Molecular Formula: C6 H10 O4 Molecular Weight (g/mol): 146.14 IUPAC Name: (2R,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol SMILES: OC[C@H]1OC=C[C@@H](O)[C@H]1O