missing translation for 'onlineSavingsMsg'
Learn More

D-Dimer Antibody (DD1), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18739272 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18739272 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18739272 Supplier Novus Biologicals Supplier No. NB1108374AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

D-Dimer Monoclonal antibody specifically detects D-Dimer in Human, Monkey, Baboon samples. It is validated for Western Blot, ELISA, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen D-Dimer
Applications Western Blot, ELISA, Immunoassay
Classification Monoclonal
Clone DD1
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunoassay
Formulation 50mM Sodium Borate
Gene Alias D Dimer
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Monkey, Baboon
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.