missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody (AT13E10), DyLight 350, Novus Biologicals™

Product Code. 18187639 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18187639 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18187639 Supplier Novus Biologicals Supplier No. NBP261163UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytosolic Sulfotransferase 2A1/SULT2A1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 2A1/SULT2A1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone AT13E10
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1
Host Species Mouse
Immunogen Recombinant human Cytosolic Sulfotransferase 2A1/SULT2A1 (1-285aa) purified from E.coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.