missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus p65 Antibody (rCMV/9827), Novus Biologicals™
Mouse Monoclonal Antibody
256.00€ - 484.00€
Specifications
| Antigen | Cytomegalovirus p65 |
|---|---|
| Clone | rCMV/9827 |
| Concentration | 0.2 mg/ml |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30291399
|
Novus Biologicals
NBP3-44629-20ug |
20 μg |
256.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30297834
|
Novus Biologicals
NBP3-44629-100ug |
100 μg |
484.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytomegalovirus p65 Monoclonal antibody specifically detects Cytomegalovirus p65 in Cytomegalovirus samples. It is validated for ELISA,ImmunohistochemistrySpecifications
| Cytomegalovirus p65 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant Cytomegalovirus p65 protein (Uniprot: P06725). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| rCMV/9827 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cytomegalovirus (CMV) | |
| CMV, p65 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title