missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin, HMW Rabbit anti-Human, Clone: KRTH/4392R, Novus Biologicals™

Rabbit Monoclonal Antibody

528.00€

Specifications

Antigen Cytokeratin, HMW
Clone KRTH/4392R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18622599
View Documents
Novus Biologicals
NBP3-07286-100ug
100 μg
528.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin, HMW Monoclonal specifically detects Cytokeratin, HMW in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Cytokeratin, HMW
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
CK-2P, cytokeratin 2, Cytokeratin-2P, HUMCYT2A, K2P, K76, keratin 76, keratin, type II cytoskeletal 2 oral, Keratin-76, KRT2Bcytokeratin-2P, KRT2Pkeratin 2p, KRT76, Type-II keratin Kb9
Synthetic peptide corresponding to Cytokeratin, HMW residues within aa300-400 of Cytokeratin, HMW
Primary
Store at 4°C.
KRTH/4392R
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
51350
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.