missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7, 8 Antibody (CAM5.2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44630-20ug
This item is not returnable.
View return policy
Description
Cytokeratin 7, 8 Monoclonal antibody specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry
Specifications
| Cytokeratin 7, 8 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| CAM5.2 | |
| Unconjugated | |
| CK7, CK-7, CK8, CK-8, Cytokeratin-7, Cytokeratin-8, keratin 8, keratin, type II cytoskeletal 7, keratin-7 | |
| Human colorectal carcinoma cell line HT29 | |
| 20 μg | |
| Cellular Signaling | |
| 3855 | |
| Store at 4⪚C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction