missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 5/6/18 Mouse anti-Human, Rat, Bovine, Clone: LP34, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | Cytokeratin 5/6/18 |
|---|---|
| Clone | LP34 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 0.1-0.2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Cytokeratin 5/6/18 Monoclonal specifically detects Cytokeratin 5/6/18 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 5/6/18 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3852 | |
| IgG1 κ | |
| Protein A or G purified |
| LP34 | |
| Immunohistochemistry-Paraffin 0.1-0.2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Bovine | |
| 58 kDa cytokeratin, CK5, CK-5, cytokeratin-5, DDD, EBS2, epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types, K5, keratin 5, keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockaynetypes), keratin, type II cytoskeletal 5, Keratin-5, KRT5A, Type-II keratin Kb5 | |
| Detergent-insoluble fraction of psoriatic human epidermis | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title