Learn More
Invitrogen™ Cytokeratin 20 Recombinant Rabbit Monoclonal Antibody (17A11)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560695
This item is not returnable.
View return policy
Description
Cytokeratin 20 Recombinant Monoclonal Antibody for ELISA, Flow, IHC (P)
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
Specifications
| Cytokeratin 20 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35900 | |
| KRT20 | |
| A synthesized peptide derived from Human KRT20. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 17A11 | |
| Unconjugated | |
| KRT20 | |
| 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54474 | |
| -20°C or -80°C if preferred | |
| Liquid |