missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody (A53-B/A2.26 + BA17), Novus Biologicals™
Mouse Monoclonal Antibody
264.00€ - 474.00€
Specifications
| Antigen | Cytokeratin 19 |
|---|---|
| Clone | A53-B/A2.26 + BA17 |
| Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18158403
|
Novus Biologicals
NBP2-34272-0.02MG |
0.02 mg |
264.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193105
|
Novus Biologicals
NBP2-34272-0.1MG |
0.1 mg |
474.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytokeratin 19 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P08727 | |
| 3880 | |
| Human breast cancer MCF-7 cells (A53-B/A2); Human mammary epithelial organoids (BA17) | |
| Primary | |
| Store at 4C. | |
| 40 kDa |
| A53-B/A2.26 + BA17 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuroscience, Stem Cell Markers | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
| KRT19 | |
| IgG2a κ, IgG1 κ | |
| Protein A purified | |
| This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40kDa. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title