missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Antibody (3G4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37468
This item is not returnable.
View return policy
Description
Cytokeratin 13 Monoclonal specifically detects Cytokeratin 13 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| Cytokeratin 13 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
| Mouse | |
| 52 kDa | |
| 0.1 mL | |
| Cytoskeleton Markers | |
| 3860 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 3G4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| KRT13 | |
| Purified recombinant fragment of human KRT13 (AA: 143-295) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction