Learn More
Cytokeratin 10 Antibody (rKRT10/6923), Novus Biologicals™
Mouse Monoclonal Antibody
249.00€ - 494.00€
Specifications
| Antigen | Cytokeratin 10 |
|---|---|
| Clone | rKRT10/6923 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652174
|
Novus Biologicals
NBP3-20243-100ug |
100 μg |
494.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660374
|
Novus Biologicals
NBP3-20243-20ug |
20 μg |
249.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 10 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10mM PBS with 0.05% BSA | |
| 3858 | |
| IgG1 κ | |
| Protein A or G purified |
| rKRT10/6923 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Recombinant fragment (around aa384-584) of human Cytokeratin 10 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |