missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Antibody (OTI4D9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
| Antigen | Cytohesin 2 |
|---|---|
| Clone | OTI4D9 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:150 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306254
|
Novus Biologicals
NBP3-44913-30ul |
30 μL |
256.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306203
|
Novus Biologicals
NBP3-44913-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytohesin 2 Monoclonal antibody specifically detects Cytohesin 2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Cytohesin 2 | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2) | |
| Full length human recombinant protein of human CYTH2 (NP_059431) produced in HEK293T cell. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI4D9 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 9266 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title