missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome P450 4A Antibody (M25-P2A10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

190.00€ - 361.00€

Specifications

Antigen Cytochrome P450 4A
Clone M25-P2A10
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Applications Western Blot, ELISA, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697247
View Documents
Novus Biologicals
NBP2-50212-0.025ml
0.025 mL
190.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18672478
View Documents
Novus Biologicals
NBP2-50212
0.1 mL
361.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytochrome P450 4A Monoclonal antibody specifically detects Cytochrome P450 4A in Human, Mouse, Rat, Drosophila, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

Cytochrome P450 4A
1 mg/mL
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Mouse
Lipid and Metabolism
PBS
1579
IgG1 κ
Protein A purified
M25-P2A10
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Monoclonal
Purified
RUO
Human, Mouse, Rat, Drosophila, Xenopus species
20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega
Peptide Sequence - KNGIHLRLR (amino acids 499 - 507)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.