missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b5 Outer Mitochondrial Membrane Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92276-0.02ml
This item is not returnable.
View return policy
Description
Cytochrome b5 Outer Mitochondrial Membrane Polyclonal antibody specifically detects Cytochrome b5 Outer Mitochondrial Membrane in Human, Mouse samples. It is validated for Western Blot
Specifications
| Cytochrome b5 Outer Mitochondrial Membrane | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CYB5M, CYB5-M, CYPB5M, Cytochrome b5 outer mitochondrial membrane isoform, cytochrome b5 type B, cytochrome b5 type B (outer mitochondrial membrane), DKFZp686M0619, OMB5, outer mitochondrial membrane cytochrome b5, type 2 cyt-b5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 16-150 of human CYB5B (NP_085056.2). KGQEVETSVTYYRLEEVAKRNSLKELWLVIHGRVYDVTRFLNEHPGGEEVLLEQAGVDASESFEDVGHSSDAREMLKQYYIGDIHPSDLKPESGSKDPSKNDTCKSCWAYWILPIIGAVLLGFLYRYYTSESKSS | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 80777 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction