missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cystatin SN Polyclonal antibody specifically detects Cystatin SN in Human samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Conjugate | mFluor Violet 500 SE |
| Gene Alias | cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Cystatin SN (Accession#: P01037-1; Met 1-Ser 141) |
| Purification Method | Antigen and protein A Affinity-purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?