missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin F Monoclonal Antibody (292103)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561056
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. This gene encodes a glycosylated cysteine protease inhibitor with a putative role in immune regulation through inhibition of a unique target in the hematopoietic system. Expression of the protein has been observed in various human cancer cell lines established from malignant tumors.
Specifications
| Cystatin F | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| O76096 | |
| CST7 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin F, Gly20-His145. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 292103 | |
| Unconjugated | |
| CST7 | |
| CMAP; CST7; cystatin 7; cystatin F; cystatin F (leukocystatin); cystatin-7; Cystatin-F; cystatin-like metastasis-associated protein; Leukocystatin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8530 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction