missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin F Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92532-0.02ml
This item is not returnable.
View return policy
Description
Cystatin F Polyclonal antibody specifically detects Cystatin F in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Cystatin F | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-145 of human CST7 (NP_003641.3). GPSPDTCSQDLNSRVKPGFPKTIKTNDPGVLQAARYSVEKFNNCTNDMFLFKESRITRALVQIVKGLKYMLEVEIGRTTCKKNQHLRLDDCDFQTNHTLKQTLSCYSEVWVVPWLQHFEVPVLRCH | |
| 0.02 mL | |
| Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
| 8530 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction