missing translation for 'onlineSavingsMsg'
Learn More

Cystatin E/M/CST6 Antibody, Novus Biologicals™

Product Code. 18771694 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18771694 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18771694 Supplier Novus Biologicals Supplier No. NBP169005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cystatin E/M/CST6 Polyclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Dilution Western Blot 1.0 ug/ml
Gene Symbols CST6
Host Species Rabbit
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.