missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4Z1 Antibody (N7-P2G5*D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 351.00€
Specifications
| Antigen | CYP4Z1 |
|---|---|
| Clone | N7-P2G5*D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641417
|
Novus Biologicals
NBP2-50215-0.025ml |
0.025 mL |
190.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636136
|
Novus Biologicals
NBP2-50215 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP4Z1 Monoclonal antibody specifically detects CYP4Z1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP4Z1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, metabolism | |
| PBS | |
| 199974 | |
| IgG1 κ | |
| Protein A purified |
| N7-P2G5*D8 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CYP4A20, CYPIVZ1, cytochrome P450 4Z1, cytochrome P450, family 4, subfamily Z, polypeptide 1, EC 1.14.14.1 | |
| Ovalbumin-conjugated synthetic peptide KLAPDHSRPP (C-terminal sequence) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title