missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CYP4V2 Polyclonal antibody specifically detects CYP4V2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | CYP4V2 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | BCD, CYP4AH1, cytochrome P450 4V2, cytochrome P450, family 4, subfamily V, polypeptide 2, EC 1.14, EC 1.14.14.1, FLJ18432, MGC43534 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200-300 of human CYP4V2 (NP_997235.3).,, Sequence:, LMFKEGWEHKKSLQILHTFTNSVIAERANEMNANEDCRGDGRGSAPSKNKRRAFLDLLLSVTDDEGNRLSHEDIREEVDTFMFEGHDTTAAAINWSLYLLGSNPEVQKKVDHELDDVFGKSDRPATVEDLKKLRYLECVIKETLRLFPSVPLFARSVSEDCEVAGYRVLKGTEAVIIPYALHRDPRYFPNPEEFQPERFFPENAQGRHPYAYVPFSAGPRNCIGQKFAVMEEKTILSCILRHFWIESNQKREELGLEGQLILRPSNGIWIKLKRRNADER |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?