missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4B1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Description
Sequence: MVPSFLSLSFSSLGLWASGLILVLGFLKLIHLLLRRQTLAKAMDKFPGPPTHWLFGHALEIQETGSLDKVVSWAHQFPYAHPLWFGQFIGFLNIYEPDYAKAVYSRGDPKAPDVYDFFLQWIGRGLLVLEGPKWLQHRKLLTPGFHYDVLKPYVAVFTESTRIMLDKWEEKAREGKSFDIFCDVGHMALNTLMKCTFGRGDTGLGHSRDSSYYLAVSDLTLLMQQRLVSFQYHNDFIYWLTPHGRRFLRACQVAHDHTDQVIRERKAALQDEKVRKKIQNRRHLDFLDILLGARDEDDIKLSDADLRAEVDTFMFEGHDTTTSGISWFLYCMALYPEHQHRCREEVREILGDQDFFQWDDLGKMTYLTMCIKESFRLYPPVPQVYRQLSKPVTFVDGRSLPAGSLISMHIYALHRNSAVWPDPEVFDSLRFSTENASKRHPFAFMPFSAGPRNCIGQQFAMSEMKVVTAMCLLRFEFSLDPSRLPIKMPQLVLRSKNGFHLHLKPLGPGSGK
Specifications
Specifications
| Antigen | CYP4B1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene | cytochrome P450, family 4, subfamily B, polypeptide 1 |
| Gene Accession No. | BC017758.1 |
| Gene Alias | CYPIVB1/P-450HP |
| Gene Symbols | CYP4B1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?