missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP26B1 Monoclonal Antibody (OTI4A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811497S
This item is not returnable.
View return policy
Description
CYP26B1 Monoclonal antibody specifically detects CYP26B1 in Human samples. It is validated for Western BlotSpecifications
| CYP26B1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00425 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10643 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4A8 | |
| Unconjugated | |
| IGF2BP3 | |
| CP26; CYP26A2; CYP26B1; Cytochrome P450 26A2; Cytochrome P450 26B1; cytochrome P450 family 26 subfamily A member 1; cytochrome P450 family 26 subfamily B member 1; cytochrome P450 retinoic acid-inactivating 2; cytochrome P450 retinoid metabolizing protein; cytochrome P450, 26, retinoic acid B1; cytochrome P450, family 26, subfamily b, polypeptide 1; cytochrome P450, subfamily XXVIB, polypeptide 1; cytochrome P450RAI-2; dol; fc21d03; P450 26A2; P450 enzyme to degrade retinoic acid; P450RAI2; P450RAI-2; retinoic acid B1; retinoic acid hydroxylase; retinoic acid-metabolizing cytochrome; RHFCA; stocksteif; wu:fc21d03; wu:fc26h10; zgc:76999 | |
| Full length human recombinant protein of human IGF2BP3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction