missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97885-50ul
This item is not returnable.
View return policy
Description
CYP1B1 Polyclonal antibody specifically detects CYP1B1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CYP1B1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase | |
| Produced in rabbits immunized with E. coli-derived Human CYP1B1 fragment. | |
| 50 μg | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1545 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction