missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, Fc Tag
457.25€ - 3344.45€
Specifications
Accession Number | XP_001106892.1 |
---|---|
Molecular Weight (g/mol) | 32.1 kDa |
Source | Cynomolgus / Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus. |
Immunogen | Met 1 - Ala 53 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15836027
|
ACROBiosystems
BCA-C5253-1MG |
1 mg |
3344.45€
1mg |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
15826027
|
ACROBiosystems
BCA-C5253-100UG |
100 μg |
457.25€
100µg |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
XP_001106892.1 | |
Cynomolgus / Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus. | |
-20°C | |
BCMA | |
Cynomolgus Monkey, Rhesus Macaque | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
32.1 kDa | |
Met 1 - Ala 53 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |