missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclosporin A Antibody (CSZ22), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2569R
This item is not returnable.
View return policy
Description
Cyclosporin A Monoclonal specifically detects Cyclosporin A in Non-species specific samples. It is validated for ELISA.
Specifications
| Cyclosporin A | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PPIA | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cyclosporin A conjugated with BSA. | |
| 0.2 mL | |
| Cancer, Cell Cycle and Replication, Stem Cell Markers | |
| 5478 | |
| Non-species specific | |
| Purified |
| ELISA | |
| CSZ22 | |
| ELISA | |
| Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognises Cyclophilin A in buffer solutions and whole blood extracts with Kd = 10^-8 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction