missing translation for 'onlineSavingsMsg'
Learn More

Cycloprate, TRC

Product Code. 30133381
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133381 250 mg 250mg
30004264 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133381 Supplier Toronto Research Chemicals Supplier No. TRCC998970250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54460-46-7
Molecular Formula C20 H38 O2
Molecular Weight (g/mol) 310.51
Synonym Hexadecyl Cyclopropanecarboxylate, ZR 856, Zardex, Cyclopropanecarboxylic Acid, Hexadecyl Ester;
IUPAC Name hexadecyl cyclopropanecarboxylate
SMILES CCCCCCCCCCCCCCCCOC(=O)C1CC1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18-22-20(21)19-16-17-19/h19H,2-18H2,1H3
Formula Weight 310.2872
Percent Purity >95%
Chemical Name or Material Cycloprate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.